Learn More
Analytical Sales & Services Vial Kit: 4mL (1 Dram) 15 x 45mm Amber Borosilicate Glass Screw Vials with Solid Teflon® Lined Screw Caps, Pkg/144
Catalog No. NC3791419
Encompass
Advantage™ Assembled Vial Kit. Contains: 4mL (1 Dram) 15 x 45mm Amber Borosilicate Glass Screw Vials with Solid Teflon® Lined Screw Caps
Catalog No. NC3791419 Supplier Analytical Sales & Services Supplier No. ADV69302A
Advantage™ Assembled Vial Kit. Contains: 4mL (1 Dram) 15 x 45mm Amber Borosilicate Glass Screw Vials with Solid Teflon® Lined Screw Caps