Learn More
Analytical Sales & Services Vial Kit: 6mL (1.5 Dram) 16 x 50mm Clear Borocilicate Glass Screw Vials with Solid Teflon Lined Screw Caps
Catalog No. 5000806157
Encompass
Advantage™ Assembled Vial Kit. Contains: 6mL (1.5 Dram) 16 x 50mm Clear Borocilicate Glass Screw Vials with Solid Teflon Lined Screw Caps
Catalog No. 50-008-06157 Supplier Analytical Sales & Services Supplier No. ADV69304
Advantage™ Assembled Vial Kit. Contains: 6mL (1.5 Dram) 16 x 50mm Clear Borocilicate Glass Screw Vials with Solid Teflon Lined Screw Caps