Learn More
Analytical Sales & Services Vial Kit: 8mL (2 Dram) 17 x 60mm Clear Borosilicate Glass Screw Vials with Solid Teflon Lined Screw Caps
Catalog No. 5000805720
Encompass
Advantage™ Assembled Vial Kit. Contains: 8mL (2 Dram) 17 x 60mm Clear Borosilicate Glass Screw Vials with Solid Teflon Lined Screw Caps
Catalog No. 50-008-05720 Supplier Analytical Sales & Services Supplier No. ADV69306
Advantage™ Assembled Vial Kit. Contains: 8mL (2 Dram) 17 x 60mm Clear Borosilicate Glass Screw Vials with Solid Teflon Lined Screw Caps