Learn More
Thermo Scientific™ Vial Kit with P/S Lined Caps
Catalog No. B78006PSSWR Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
B7800-6PSSWR Clear
1 options
Catalog No. B7800-6PSSWR Supplier Thermo Scientific™ Supplier No. B78006PS
Get an Extra Case of Product with Purchase
View all promotions
Add to Cart
Add to Cart
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Storage vials range from 2mL to 40mL.
  • Clear vial
  • Polyseal-lined solid-top storage caps
  • Not autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Material (Liner) Polyseal™
Color Clear
Volume (Metric) 40 mL
Quantity 100/Pk.