Learn More
Thermo Scientific™ Vial Kit with Polyseal Caps
Catalog No. 03377223
Change view
Click to view available options
Color:
Clear
Catalog No. Color
03-377-223 Clear
1 options
This item is not returnable. View return policy
Catalog No. 03-377-223 Supplier Thermo Scientific™ Supplier No. B78012PS
This item is not returnable. View return policy
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample.
  • Clear vial
  • PTFE-lined solid-top storage caps
  • PTFE on film/foam backing offers broad chemical resistance
  • Not autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Material (Liner) PTFE/Silicone
Color Clear
Volume (Metric) 4 mL
Quantity 100/Pk.