Learn More
Chemglass Life Sciences Vial, 2.0mL, Clear with White Grad Spot, Large Opening, 12x32mm, 11mm Crimp
SDP
Catalog No. 50230755
Encompass_Preferred
Vial, 2.0mL, Clear with White Grad Spot, Large Opening, 12x32mm, 11mm Crimp. The 40% larger opening prevents broken needles due to the increased target area. The neck finish allows for the use of the patented Poly Crimp™ seals or standard aluminum seals. This vial is designed to work auto samples incorporating a robotic arm as well as other auto samplers. Clear. The vial incorporates the unique Step Vial design that precisely centers a limited volume insert in the vial neck. Available with or without graduated marking spots.
Catalog No. 50-230-755 Supplier Chemglass Life Sciences Supplier No. CV11871232
Vial, 2.0mL, Clear with White Grad Spot, Large Opening, 12x32mm, 11mm Crimp. The 40% larger opening prevents broken needles due to the increased target area. The neck finish allows for the use of the patented Poly Crimp™ seals or standard aluminum seals. This vial is designed to work auto samples incorporating a robotic arm as well as other auto samplers. Clear. The vial incorporates the unique Step Vial design that precisely centers a limited volume insert in the vial neck. Available with or without graduated marking spots.