Learn More
Chemglass Life Sciences Vial, 2.0mL, Amber with White Grad Spot, Large Opening, 12x32mm, GPI 10-425
SDP
Catalog No. 50230744
Encompass_Preferred
Vial, 2.0mL, Amber with White Grad Spot, Large Opening, 12x32mm, GPI 10-425. With a 40% larger opening, the Large Opening Vial is specifically designed to eliminate broken needles due to the increased target area. Amber borosilicate. These vials incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial. Available with a graduated marking spot. Vials available silanized.
Catalog No. 50-230-744 Supplier Chemglass Life Sciences Supplier No. CV11631232
Vial, 2.0mL, Amber with White Grad Spot, Large Opening, 12x32mm, GPI 10-425. With a 40% larger opening, the Large Opening Vial is specifically designed to eliminate broken needles due to the increased target area. Amber borosilicate. These vials incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial. Available with a graduated marking spot. Vials available silanized.