Learn More
Chemglass Life Sciences 1mL Clear High Recovery Vial, V Bottom, Micro Product, Graduated, 13-425 Black Phenolic Open Top Cap and Red PTFE/Silicone Septa, 13mm OD x 41mm Height
SDP
Catalog No. 50143870
Encompass_Preferred
Vial, Micro Product, 1mL, GPI 13-425. Vials are manufactured from Type 1, Class A Borosilicate glass and come with an open-top cap with PTFE Silicone septa. Vials come with a fire glazed bottom to eliminate particulate unlike other micro vials with ground bottoms. These vials have a conical interior bottom that allows for maximum retrieval with a syringe or micropipette. Vials can be used for small-scale reactions, centrifugation and storage. Sold in cases of 12.
Catalog No. 50-143-870 Supplier Chemglass Life Sciences Supplier No. CG4913001
Vial, Micro Product, 1mL, GPI 13-425. Vials are manufactured from Type 1, Class A Borosilicate glass and come with an open-top cap with PTFE Silicone septa. Vials come with a fire glazed bottom to eliminate particulate unlike other micro vials with ground bottoms. These vials have a conical interior bottom that allows for maximum retrieval with a syringe or micropipette. Vials can be used for small-scale reactions, centrifugation and storage. Sold in cases of 12.