Learn More
Chemglass Life Sciences 3mL Clear High Recovery Vial, V Bottom, Micro Product, Graduated, 20-400 Black Phenolic Open Top Cap and Gray PTFE/Silicone Septa, 20mm OD x 47mm Height
SDP
Catalog No. 50143891
Encompass_Preferred
Vial, Micro Product, 3mL, GPI 20-400. Vials are manufactured from Type 1, Class A Borosilicate glass and come with an open-top cap with PTFE Silicone septa. Vials come with a fire glazed bottom to eliminate particulate unlike other micro vials with ground bottoms. These vials have a conical interior bottom that allows for maximum retrieval with a syringe or micropipette. Vials can be used for small-scale reactions, centrifugation and storage. Sold in cases of 12.
Catalog No. 50-143-891 Supplier Chemglass Life Sciences Supplier No. CG4913003
Vial, Micro Product, 3mL, GPI 20-400. Vials are manufactured from Type 1, Class A Borosilicate glass and come with an open-top cap with PTFE Silicone septa. Vials come with a fire glazed bottom to eliminate particulate unlike other micro vials with ground bottoms. These vials have a conical interior bottom that allows for maximum retrieval with a syringe or micropipette. Vials can be used for small-scale reactions, centrifugation and storage. Sold in cases of 12.