Learn More
Macherey-Nagel Screw neck vial N9, Micro-vial, clear with integrated 0.2 mL insert
Catalog No. NC1555939
Encompass_Preferred
Screw Neck Vial N 9 - outer diameter: 11.6 mm, outer height: 32 mm - clear, flat bottom, with integrated Insert 0.2 mL, conical - pack of 100
Catalog No. NC1555939 Supplier Macherey-Nagel Supplier No. 702007
Screw Neck Vial N 9 - outer diameter: 11.6 mm, outer height: 32 mm - clear, flat bottom, with integrated Insert 0.2 mL, conical - pack of 100