Learn More
Trajan Scientific La-Pha-Pack , Vial, ND22, 3rd Hyrdolytic Glass, Snap, 15mL, 48x26mm, 100 Pack. Made in Germany.
Catalog No. 501037388
Encompass_Preferred
La-Pha-Pack , Vial, ND22, 3rd Hyrdolytic Glass, Snap, 15mL, 48x26mm, 100 Pack. Made in Germany.
Catalog No. 50-103-7388 Supplier Trajan Scientific Supplier No. LAP.20090784
La-Pha-Pack , Vial, ND22, 3rd Hyrdolytic Glass, Snap, 15mL, 48x26mm, 100 Pack. Made in Germany.