Learn More
Chemglass Life Sciences Vial Only, 1.0mL, Amber, Glass, WISP Shell, 8x40mm
SDP
Catalog No. 501532505
Encompass_Preferred
Vial, 0.8mL, 8x40mm, Amber, Flat Bottom. The 8x30mm vial is primarily used with the 60 and 100 trays for the Perkin-Elmer autosamplers. The 8x40mm vials are primarily used in the Al, Carlo Erba, Dionex and Dynatech autosamplers. Vials are manufactured from Type 1, Class A borosilicate glass.
Catalog No. 50-153-2505 Supplier Chemglass Life Sciences Supplier No. CV21040840
Vial, 0.8mL, 8x40mm, Amber, Flat Bottom. The 8x30mm vial is primarily used with the 60 and 100 trays for the Perkin-Elmer autosamplers. The 8x40mm vials are primarily used in the Al, Carlo Erba, Dionex and Dynatech autosamplers. Vials are manufactured from Type 1, Class A borosilicate glass.