Learn More
Chemglass Life Sciences Vial Only, 1.0mL, Clear, Glass, WISP Shell, 8x40mm
SDP
Catalog No. 50230872
Encompass_Preferred
Vial Only, 1.0mL, Clear, Glass, WISP Shell, 8x40mm. This vial is designed for use in the Waters WISP 96-Position Auto Sampler. Available in clear and amber borosilicate or polypropylene for easy disposal. These vials feature thicker walls for safer sample handling. The limited volume shell vials have a precisely designed conical bottom which permits maximum sample evacuation.
Catalog No. 50-230-872 Supplier Chemglass Life Sciences Supplier No. CV21030840
Vial Only, 1.0mL, Clear, Glass, WISP Shell, 8x40mm. This vial is designed for use in the Waters WISP 96-Position Auto Sampler. Available in clear and amber borosilicate or polypropylene for easy disposal. These vials feature thicker walls for safer sample handling. The limited volume shell vials have a precisely designed conical bottom which permits maximum sample evacuation.