Learn More
Chemglass Life Sciences Vial Only, Sample, 20mL, Clear, 28x57mm, GPI 24-400
SDP
Catalog No. 50230654
Encompass_Preferred
Vial Only, Sample, 20mL, Clear, 28x57mm, GPI 24-400. Clear sample vials packaged in convenient LAB-PACS. Made from Type 1 borosilicate glass. Vials are conveniently packed in shrink-wrapped packs of 100. Caps sold separately.
Catalog No. 50-230-654 Supplier Chemglass Life Sciences Supplier No. CG490206
Vial Only, Sample, 20mL, Clear, 28x57mm, GPI 24-400. Clear sample vials packaged in convenient LAB-PACS. Made from Type 1 borosilicate glass. Vials are conveniently packed in shrink-wrapped packs of 100. Caps sold separately.