Learn More
Chemglass Life Sciences Vial Only, Sample, 4mL, Amber, 15x45mm, GPI 13-425
SDP
Catalog No. 501531777
Encompass_Preferred
Vial Only, Sample, 4mL, Amber, 15x45mm, GPI 13-425. Amber sample vials packaged in convenient LAB-PACS. Made from Type 1 borosilicate glass. Vials are conveniently packed in shrink-wrapped packs of 100.
Catalog No. 50-153-1777 Supplier Chemglass Life Sciences Supplier No. CG490602
Vial Only, Sample, 4mL, Amber, 15x45mm, GPI 13-425. Amber sample vials packaged in convenient LAB-PACS. Made from Type 1 borosilicate glass. Vials are conveniently packed in shrink-wrapped packs of 100.