Learn More
Chemglass Life Sciences Vial Only, Sample, 8mL, Clear, 17x60mm, GPI 15-425
SDP
Catalog No. 50230651
Encompass_Preferred
Vial Only, Sample, 8mL, Clear, 17x60mm, GPI 15-425. Clear sample vials packaged in convenient LAB-PACS. Made from Type 1 borosilicate glass. Vials are conveniently packed in shrink-wrapped packs of 100. Caps sold separately.
Catalog No. 50-230-651 Supplier Chemglass Life Sciences Supplier No. CG490203
Vial Only, Sample, 8mL, Clear, 17x60mm, GPI 15-425. Clear sample vials packaged in convenient LAB-PACS. Made from Type 1 borosilicate glass. Vials are conveniently packed in shrink-wrapped packs of 100. Caps sold separately.