Learn More
Chemglass Life Sciences Vial Only, Sample, 8mL, Amber, 17x60mm, GPI 15-425
SDP
Catalog No. 501531778
Encompass_Preferred
Vial Only, Sample, 8mL, Amber, 17x60mm, GPI 15-425. Amber sample vials packaged in convenient LAB-PACS. Made from Type 1 borosilicate glass. Vials are conveniently packed in shrink-wrapped packs of 100.
Catalog No. 50-153-1778 Supplier Chemglass Life Sciences Supplier No. CG490603
Vial Only, Sample, 8mL, Amber, 17x60mm, GPI 15-425. Amber sample vials packaged in convenient LAB-PACS. Made from Type 1 borosilicate glass. Vials are conveniently packed in shrink-wrapped packs of 100.