Learn More
Chemglass Life Sciences Vial, 2.0mL, Clear, Glass, Shell, Round Bottom, 12x32mm
SDP
Catalog No. 50230879
Encompass_Preferred
Vial, 2.0mL, Clear, Glass, Shell, Round Bottom, 12x32mm. Shell vials are the most economical alternative for use in an auto sampler. Feature thicker walls for safer sample handling.Require Snap Plugs
Catalog No. 50-230-879 Supplier Chemglass Life Sciences Supplier No. CV21101232
Vial, 2.0mL, Clear, Glass, Shell, Round Bottom, 12x32mm. Shell vials are the most economical alternative for use in an auto sampler. Feature thicker walls for safer sample handling.Require Snap Plugs