Learn More
Microsolv Technology Corporation Vial Pe Plug Clear Shellvia
Catalog No. 5001451997
Encompass_Preferred
Vial & Cap Kit includes 100 EA 2ml, Push Top, Clear Autosampler Vials and 100 EA matching, translucent, polyethylene Push Caps for Micro-Insert. MicroSolv Brand Easy Purchase Pack. 1/PK.
Catalog No. 50-014-51997 Supplier Microsolv Technology Corporation Supplier No. 9502CCSVEPP
Vial & Cap Kit includes 100 EA 2ml, Push Top, Clear Autosampler Vials and 100 EA matching, translucent, polyethylene Push Caps for Micro-Insert. MicroSolv Brand Easy Purchase Pack. 1/PK.