Learn More
Electron Microscopy Sciences Polyethylene Vial 1 ml, 9 mm diameter, 32 mm height
SDP
Supplier:  Electron Microscopy Sciences 7262112
Encompass_Preferred
Sample vials with a captive snap closure; flat bottom cylindrical vials with a friction-fit, flip-top closure which can not be lost. These vials are unbreakable and can withstand temperatures up to 90 degree C. The wall thickness is approx. 1 mm and these vials are ideal for use as a specimen container for the storage of small samples, or as embedding capsules.
Diameter: 9 mm
Height: 32 mm
Catalog No. 50-189-9958