Learn More
Electron Microscopy Sciences Polyethylene Vial 5 ml, 15 mm diameter, 50 mm height
SDP
Catalog No. 501899960
Encompass_Preferred
Sample vials with a captive snap closure; flat bottom cylindrical vials with a friction-fit, flip-top closure which can not be lost. These vials are unbreakable and can withstand temperatures up to 90 degree C. The wall thickness is approx. 1 mm and these vials are ideal for use as a specimen container for the storage of small samples, or as embedding capsules.
Diameter: 15 mm
Height: 50 mm
Catalog No. 50-189-9960 Supplier Electron Microscopy Sciences Supplier No. 7262132
Sample vials with a captive snap closure; flat bottom cylindrical vials with a friction-fit, flip-top closure which can not be lost. These vials are unbreakable and can withstand temperatures up to 90 degree C. The wall thickness is approx. 1 mm and these vials are ideal for use as a specimen container for the storage of small samples, or as embedding capsules.
Diameter: 15 mm
Height: 50 mm