Learn More
Waters Corp Autosampler Vial, Polypropylene, 12 x 32 mm, 250 µL Volume, Screw Neck
Catalog No. 50819160 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Clean polypropylene-molded vial, preferred in applications where there is a concern that compound or molecule may stick to a polar glass surface. This vial is a cost effective alternative to glass.
  • Closure type: Screw neck
  • Dimensions: 12 x 32 mm
  • Material: Polypropylene
  • Volume capacity: 250 µL
  • Type: Autosampler vial
  • Includes: Vial
  • Units per package: 100 pk
  • Application: Sample preparation
Catalog No. 50-819-160 Supplier Waters Corp Supplier No. WAT094172
Clean polypropylene-molded vial, preferred in applications where there is a concern that compound or molecule may stick to a polar glass surface. This vial is a cost effective alternative to glass.
  • Closure type: Screw neck
  • Dimensions: 12 x 32 mm
  • Material: Polypropylene
  • Volume capacity: 250 µL
  • Type: Autosampler vial
  • Includes: Vial
  • Units per package: 100 pk
  • Application: Sample preparation