Learn More
JG FINNERAN 300µL Polypropylene Limited Volume Vial, 1000/case
Catalog No. NC1705610
Encompass_Preferred
Polypropylene is translucent, compatible with many solvents and an excellent choice for storing pH sensitive samples. Maximum temperature usage is 135ºC/275ºF.
Catalog No. NC1705610 Supplier JG FINNERAN Supplier No. 30111P1232
Polypropylene is translucent, compatible with many solvents and an excellent choice for storing pH sensitive samples. Maximum temperature usage is 135ºC/275ºF.