Learn More
Chemglass Life Sciences 20-40mL Vial Rack, 50-Place, Polypropylene
SDP
Catalog No. 501941131
Encompass_Preferred
20-40mL Vial Rack, 50-Place, Polypropylene. Blue Polypropylene Heavy-Duty racks have excellent chemical resistance and can be autoclaved. Ideal for use when processing or transporting samples. Racks have an Alphanumeric grid for identification. The racks are stackable when not in use. The rack measures: 13” x 6-7/8" x 1-3/16" High, and 50 Place/Capacity, 1.1" (28mm) diameter x .920 " (23mm) deep. The vial cavities have a center drain hole 6.5mm in diameter.
Catalog No. 50-194-1131 Supplier Chemglass Life Sciences Supplier No. CG4900R10
20-40mL Vial Rack, 50-Place, Polypropylene. Blue Polypropylene Heavy-Duty racks have excellent chemical resistance and can be autoclaved. Ideal for use when processing or transporting samples. Racks have an Alphanumeric grid for identification. The racks are stackable when not in use. The rack measures: 13” x 6-7/8" x 1-3/16" High, and 50 Place/Capacity, 1.1" (28mm) diameter x .920 " (23mm) deep. The vial cavities have a center drain hole 6.5mm in diameter.