Learn More
Microsolv Technology Corporation Vial Racks, for 12x32mm, 2ml autosampler vials. Polypropylene, blue, with 50 wells that are indexed. Stackable. 5/PK.
Catalog No. 502271395
Encompass_Preferred
Convenient and easy management for transportation or storage of standard, 12mm wide autosampler vials that are used in HPLC, LCMS, GC or GCMS. This polypropylene rack is chemically resistant to most HPLC solvents. Racks are stackable for storing them. Conical and flat bottom vials can be accommodated in this rack
Catalog No. 50-227-1395 Supplier Microsolv Technology Corporation Supplier No. 9502R02BK
Convenient and easy management for transportation or storage of standard, 12mm wide autosampler vials that are used in HPLC, LCMS, GC or GCMS. This polypropylene rack is chemically resistant to most HPLC solvents. Racks are stackable for storing them. Conical and flat bottom vials can be accommodated in this rack