Learn More
Microsolv Technology Corporation Vial Rack, for 15x45mm Vials. Acrylic, clear, with 40 cavities. 1 EA.
Catalog No. 502271394
Encompass_Preferred
For easy handling and transportation of 4ml, 15x45mm autosampler vials. The clear materials makes filling easier with greater visibility compared to opaque racks. Also, racks are stackable due to the silicone feet on the base. More stable than other racks on the market due to the solid, one piece construction. Flat bottom vials can be accommodated in this 175.8mm wide x 115.5mm deep and 20mm high rack. Wells are 15.1mm in diameter.
This item is not returnable. View return policy
Catalog No. 50-227-1394 Supplier Microsolv Technology Corporation Supplier No. 9504R01
For easy handling and transportation of 4ml, 15x45mm autosampler vials. The clear materials makes filling easier with greater visibility compared to opaque racks. Also, racks are stackable due to the silicone feet on the base. More stable than other racks on the market due to the solid, one piece construction. Flat bottom vials can be accommodated in this 175.8mm wide x 115.5mm deep and 20mm high rack. Wells are 15.1mm in diameter.