Learn More
Microsolv Technology Corporation Vial Rack, for 8mm Specialty Vials. Acrylic, clear, with 50 cavities. 1 EA.
Catalog No. 502270349
Encompass_Preferred
For easy handling and transportation of 8mm wide autosampler vials. The clear materials makes filling easier with greater visibility compared to opaque racks. Also, racks are stackable due to the silicone feet on the base. More stable than other racks on the market due to the solid, one piece construction. Conical and flat bottom vials can be accommodated in this 173mm wide x 95mm deep and 20mm high rack. Wells are 8.5/3mm in diameter.
This item is not returnable. View return policy
Catalog No. 50-227-0349 Supplier Microsolv Technology Corporation Supplier No. 9508R01
For easy handling and transportation of 8mm wide autosampler vials. The clear materials makes filling easier with greater visibility compared to opaque racks. Also, racks are stackable due to the silicone feet on the base. More stable than other racks on the market due to the solid, one piece construction. Conical and flat bottom vials can be accommodated in this 173mm wide x 95mm deep and 20mm high rack. Wells are 8.5/3mm in diameter.