Learn More
Chemglass Life Sciences Storage Vial, 4mL, Complete, Clear, 15 x 45mm, 13-425 Green Open Top Cap, TFE Septa
SDP
Catalog No. 50143879
Encompass_Preferred
Storage/Reaction Vial, 4mL, Complete, Clear, 15 x 45mm, 13-425 Green Open Top Cap, TFE Septa. For reaction or storage applications, these vials are packaged in a SBS coated white tray complete with SBS partition. Boxes are marked with alpha-numeric coordinates for fast and easy vial identification. Caps are supplied installed on the vials and all sizes have the TFE lined silicone SURE-LINK septa. The 4mL is a 1 dram.
Catalog No. 50-143-879 Supplier Chemglass Life Sciences Supplier No. CG490904
Storage/Reaction Vial, 4mL, Complete, Clear, 15 x 45mm, 13-425 Green Open Top Cap, TFE Septa. For reaction or storage applications, these vials are packaged in a SBS coated white tray complete with SBS partition. Boxes are marked with alpha-numeric coordinates for fast and easy vial identification. Caps are supplied installed on the vials and all sizes have the TFE lined silicone SURE-LINK septa. The 4mL is a 1 dram.