Learn More
Grainger Vial .14 oz. 4mL PK144
Catalog No. NC9878337
Encompass_Preferred
Clear Screw Thread Vial, 4mL, Available in Clear or Amber Borosilicate Glass. Available in Clear or Amber borosilicate glass. Vials resist chemicals and permeation of oxygen. Caps have a PTFE lining backed with polypropylene foam
Catalog No. NC9878337 Supplier Grainger Supplier No. 3LEH1
Clear Screw Thread Vial, 4mL, Available in Clear or Amber Borosilicate Glass. Available in Clear or Amber borosilicate glass. Vials resist chemicals and permeation of oxygen. Caps have a PTFE lining backed with polypropylene foam