Learn More
Chemglass Life Sciences Convenience Pack, 2.0mL Amber Glass 12x32mm Shell Vial, Snap Plug
SDP
Catalog No. 501532671
Encompass_Preferred
Convenience Pack, 2.0mL Amber Glass 12x32mm Shell Vial, Snap Plug. Shell vials are the most economical alternative for use in an auto sampler. Feature thicker walls for safer sample handling.
Catalog No. 50-153-2671 Supplier Chemglass Life Sciences Supplier No. CV70551232
Convenience Pack, 2.0mL Amber Glass 12x32mm Shell Vial, Snap Plug. Shell vials are the most economical alternative for use in an auto sampler. Feature thicker walls for safer sample handling.