Learn More
Chemglass Life Sciences Vial, 2.0mL, Amber with White Grad Spot, Snap Ring, 12x32mm, 11mm Crimp
SDP
Catalog No. 501532463
Encompass_Preferred
Vial, 2.0mL, Amber with White Grad Spot, Snap Ring, 12x32mm, 11mm Crimp. The neck finish of the Snap Ring vial allows the use of Chem Snap Caps or standard aluminum seals. The Snap Ring finish eliminates the need for crimping or decapping. Designed to work with robotic arm or other auto samplers. This vial also incorporates the unique Step Vial design that precisely centers a limited volume insert in the vial neck.
Catalog No. 50-153-2463 Supplier Chemglass Life Sciences Supplier No. CV12031232
Vial, 2.0mL, Amber with White Grad Spot, Snap Ring, 12x32mm, 11mm Crimp. The neck finish of the Snap Ring vial allows the use of Chem Snap Caps or standard aluminum seals. The Snap Ring finish eliminates the need for crimping or decapping. Designed to work with robotic arm or other auto samplers. This vial also incorporates the unique Step Vial design that precisely centers a limited volume insert in the vial neck.