Learn More
Chemglass Life Sciences Vial, 2.0mL, Clear, Snap Ring, 12x32mm, 11mm Crimp
SDP
Catalog No. 501532460
Encompass_Preferred
Vial, 2.0mL, Clear, Snap Ring, 12x32mm, 11mm Crimp. The neck finish of the Snap Ring vial allows the use of Chem Snap Caps or standard aluminum seals. The Snap Ring finish eliminates the need for crimping or decapping. Designed to work with robotic arm or other auto samplers. This vial also incorporates the unique Step Vial design that precisely centers a limited volume insert in the vial neck.
Catalog No. 50-153-2460 Supplier Chemglass Life Sciences Supplier No. CV12001232
Vial, 2.0mL, Clear, Snap Ring, 12x32mm, 11mm Crimp. The neck finish of the Snap Ring vial allows the use of Chem Snap Caps or standard aluminum seals. The Snap Ring finish eliminates the need for crimping or decapping. Designed to work with robotic arm or other auto samplers. This vial also incorporates the unique Step Vial design that precisely centers a limited volume insert in the vial neck.