Learn More
Chemglass Life Sciences Vial, 2.0mL, Clear with White Grad Spot, Standard Opening, 12x32mm, 11mm Crimp
SDP
Catalog No. 50230749
Encompass_Preferred
Vial, 2.0mL, Clear with White Grad Spot, Standard Opening, 12x32mm, 11mm Crimp. The neck finish allows for the use of the patented Poly Crimp™ Seals or standard aluminum seals. This vial is designed to work auto samples incorporating a robotic arm as well as other auto samplers. Clear. The vial incorporates the unique Step Vial design that precisely centers a limited volume insert in the vial neck. Available with or without graduated marking spots. Vials available silanized.
Catalog No. 50-230-749 Supplier Chemglass Life Sciences Supplier No. CV11701232
Vial, 2.0mL, Clear with White Grad Spot, Standard Opening, 12x32mm, 11mm Crimp. The neck finish allows for the use of the patented Poly Crimp™ Seals or standard aluminum seals. This vial is designed to work auto samples incorporating a robotic arm as well as other auto samplers. Clear. The vial incorporates the unique Step Vial design that precisely centers a limited volume insert in the vial neck. Available with or without graduated marking spots. Vials available silanized.