Learn More
Chemglass Life Sciences Vial, 2.0mL, Amber, Standard Opening, 12x32mm, 11mm Crimp
SDP
Catalog No. 50230748
Encompass_Preferred
Vial, 2.0mL, Amber, Standard Opening, 12x32mm, 11mm Crimp. The neck finish allows for the use of the patented Poly Crimp™ Seals or standard aluminum seals. This vial is designed to work auto samples incorporating a robotic arm as well as other auto samplers. Amber borosilicate. The vial incorporates the unique Step Vial design that precisely centers a limited volume insert in the vial neck. Available with or without graduated marking spots. Vials available silanized.
Catalog No. 50-230-748 Supplier Chemglass Life Sciences Supplier No. CV11691232
Vial, 2.0mL, Amber, Standard Opening, 12x32mm, 11mm Crimp. The neck finish allows for the use of the patented Poly Crimp™ Seals or standard aluminum seals. This vial is designed to work auto samples incorporating a robotic arm as well as other auto samplers. Amber borosilicate. The vial incorporates the unique Step Vial design that precisely centers a limited volume insert in the vial neck. Available with or without graduated marking spots. Vials available silanized.