Learn More
Electron Microscopy Sciences Sample Storage Set Poly-Seal Screw Cap Series, 4 dram, 21 x 70 mm
SDP
Catalog No. 5028433
Encompass_Preferred
Threaded glass vials with Poly-seal caps and a cone shaped polyethylene liner. Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Tray: 9 1/4"(L) x 4 3/4"(W) x 3 1/4"(H).
Volume: 4 dram
Size: 21 x 70 mm
Catalog No. 50-284-33 Supplier Electron Microscopy Sciences Supplier No. 64270
Threaded glass vials with Poly-seal caps and a cone shaped polyethylene liner. Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Tray: 9 1/4"(L) x 4 3/4"(W) x 3 1/4"(H).
Volume: 4 dram
Size: 21 x 70 mm