Learn More
Electron Microscopy Sciences Sample Storage Set Poly-Seal Screw Cap Series, 8 dram, 25 x 95 mm
SDP
Catalog No. 5028434
Encompass_Preferred
Threaded glass vials with Poly-seal caps and a cone shaped polyethylene liner. Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Tray: 10.5"(L) x 5.5"(W) x 4.5"(H).
Volume: 8 dram
Size: 25 x 95 mm
Catalog No. 50-284-34 Supplier Electron Microscopy Sciences Supplier No. 64271
Threaded glass vials with Poly-seal caps and a cone shaped polyethylene liner. Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Tray: 10.5"(L) x 5.5"(W) x 4.5"(H).
Volume: 8 dram
Size: 25 x 95 mm