Learn More
Brookfield Engineer Labs VIAL TESTING ASSAY KIT 10PK
Catalog No. NC2706602
Encompass
DescriptionGlass Sample bottles with metal cap and septa. Compatible with Vapor Pro instruments CT-3100(-L) and CT-4200XL. Includes 10 glass sample bottles.
Catalog No. NC2706602 Supplier Brookfield Engineer Labs Supplier No. Y9900226
DescriptionGlass Sample bottles with metal cap and septa. Compatible with Vapor Pro instruments CT-3100(-L) and CT-4200XL. Includes 10 glass sample bottles.