Learn More
Molecular Dimensions VIAL TONGS (45)
SDP
Catalog No. 501038602
Encompass
Vial Tongs (45 deg.) For transfer of vials to and from cryocanes and general cryomanipulation of cryocooled crystals.
Catalog No. 50-103-8602 Supplier Molecular Dimensions Supplier No. MD7516
Vial Tongs (45 deg.) For transfer of vials to and from cryocanes and general cryomanipulation of cryocooled crystals.