Learn More
Scientific Specialties Service Inc 22mL Clear Borosilicate Screw Thread Vials with sci/spec Teflon Foam
SDP
Catalog No. NC1699883
Encompass_Preferred
22mL Clear Borosilicate Screw Thread Vials with sci/spec Teflon Foam. 6 drams. 23 x 85mm. Cap size: 20-400
Catalog No. NC1699883 Supplier Scientific Specialties Service Inc Supplier No. B69314
22mL Clear Borosilicate Screw Thread Vials with sci/spec Teflon Foam. 6 drams. 23 x 85mm. Cap size: 20-400