Learn More
Waters Corp Waters Corporation Vial, LCGC Certified, Clear Glass, 12 x 32 mm, 2 mL
Supplier:  Waters Corp WAT270946C
Encompass_Preferred
These certified vials are tested for cleanliness by HPLC and held to the tightest dimensional tolerances in the industry, ensuring reliable performance for sample preparation.
  • Clear glass material
  • 12 x 32 mm dimensions
  • 2 mL volume capacity
  • LCGC certified
  • Screw neck closure
  • Black cap
  • PTFE/Silicone seal material
  • Autosampler vial type
  • Includes vial, cap, and septum
  • 100 units per package
Catalog No. 50-818-631