Learn More
Waters Corp Waters Vial, Clear Glass, Screw Neck, 4 mL, Cap and PTFE Septum
Catalog No. 50785894 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This clear glass screw neck vial measures 15 x 45 mm and has a 4 mL volume capacity. It includes a cap and a PTFE septum and is designed as an autosampler vial for sample preparation applications.
  • Measures 15 x 45 mm
  • Provides 4 mL volume capacity
  • Features a screw neck closure type
  • Constructed from clear glass
  • Includes vial, cap, and septum
  • Suitable for sample preparation
  • Designed as an autosampler vial
  • Available in packages of 1440 units
Catalog No. 50-785-894 Supplier Waters Corp Supplier No. WAT073018
This clear glass screw neck vial measures 15 x 45 mm and has a 4 mL volume capacity. It includes a cap and a PTFE septum and is designed as an autosampler vial for sample preparation applications.
  • Measures 15 x 45 mm
  • Provides 4 mL volume capacity
  • Features a screw neck closure type
  • Constructed from clear glass
  • Includes vial, cap, and septum
  • Suitable for sample preparation
  • Designed as an autosampler vial
  • Available in packages of 1440 units