Learn More
Chemglass Life Sciences Vial, 10mL, Clear, 20mm Headspace, Round Bottom, 23x46mm, 20mm Crimp, for use with LEAP Autosamplers
SDP
Catalog No. 501532470
Encompass_Preferred
Vial, 10mL, Clear, 20mm Headspace, Round Bottom, 23x46mm, 20mm Crimp, for use with LEAP Autosamplers. These vials are manufactured from Type 1 Class A Borosilicate glass. Please Note: Pressure Release Aluminum Seals are Designed to Open When the Internal Pressure Exceeds 3.0 +/- 0.5 Bar.
Catalog No. 50-153-2470 Supplier Chemglass Life Sciences Supplier No. CV12812346
Vial, 10mL, Clear, 20mm Headspace, Round Bottom, 23x46mm, 20mm Crimp, for use with LEAP Autosamplers. These vials are manufactured from Type 1 Class A Borosilicate glass. Please Note: Pressure Release Aluminum Seals are Designed to Open When the Internal Pressure Exceeds 3.0 +/- 0.5 Bar.