Learn More
Chemglass Life Sciences Vial, 10mL, Clear, 20mm Headspace, Flat Bottom, 23x46mm, 20mm Crimp
SDP
Catalog No. 501532467
Encompass_Preferred
Vial, 10mL, Clear, 20mm Headspace, Flat Bottom, 23x46mm, 20mm Crimp. These vials are manufactured from Type 1 Class A Borosilicate glass. The standard flat bottom vials can be used with Hewlett-Packard, Carlo Erba and Shimadzu. Please Note: Pressure Release Aluminum Seals are Designed to Open When the Internal Pressure Exceeds 3.0 +/- 0.5 Bar.
Catalog No. 50-153-2467 Supplier Chemglass Life Sciences Supplier No. CV12802346
Vial, 10mL, Clear, 20mm Headspace, Flat Bottom, 23x46mm, 20mm Crimp. These vials are manufactured from Type 1 Class A Borosilicate glass. The standard flat bottom vials can be used with Hewlett-Packard, Carlo Erba and Shimadzu. Please Note: Pressure Release Aluminum Seals are Designed to Open When the Internal Pressure Exceeds 3.0 +/- 0.5 Bar.