Learn More
Thomas Scientific 2D Cryovials, 2mL, Self Standing, External Threading, Sterile
SDP
Catalog No. 501461720
Encompass
PolarSafe™ 2D tubes are the ideal consumable for rapid processing of biological samples. The embedded Datamatix code inserted in the bottom of each tube corresponds to the “Code 128” barcode featured on the body of the tube. They provide time-saving and accurate organization and tracking of valuable samples. Innovative “channels of evacuation” designed between the tube and the 2D chip allows fluid to easily drain The 2D tubes are offered in both 2 and 5mL sizes Available with internally or externally threaded caps
Catalog No. 50-146-1720 Supplier Thomas Scientific Supplier No. 1186P25
PolarSafe™ 2D tubes are the ideal consumable for rapid processing of biological samples. The embedded Datamatix code inserted in the bottom of each tube corresponds to the “Code 128” barcode featured on the body of the tube. They provide time-saving and accurate organization and tracking of valuable samples. Innovative “channels of evacuation” designed between the tube and the 2D chip allows fluid to easily drain The 2D tubes are offered in both 2 and 5mL sizes Available with internally or externally threaded caps