Learn More
Lonza Walkersville ViaLight Plus Cell Proliferation and Cytotoxicity BioAssay Kit, 500 Test
Supplier:  Lonza Walkersville LT07221
Encompass
ViaLight Plus BioAssay Kits are designed to provide unprecedented speed and sensitivity for cytotoxicity and cell proliferation studies. The ViaLight Assay incorporates bioluminescent detection of cellular ATP as a measure of viability. Measurement of ATP is the most accurate, effective, and direct way of determining the number of living cells in culture.
Catalog No. NC1985481