Learn More
TARGETMOL CHEMICALS INC VIALININ A 5MG
SDP
Catalog No. 502580972
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Vialinin A (Terrestrin A) (Terrestrin A) is a p-terphenyl compound with antioxidant properties [1]. Vialinin A is a potent inhibitor of TNF-(alpha) USP4 USP5 and sentrin/SUMO-specific protease 1 (SENP1). Vialinin A (Terrestrin A) is able to be used for autoimmune diseases and cancer research [2] [3]. purity: 99%
Catalog No. 50-258-0972 Supplier TARGETMOL CHEMICALS INC Supplier No. T221685MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Vialinin A (Terrestrin A) (Terrestrin A) is a p-terphenyl compound with antioxidant properties [1]. Vialinin A is a potent inhibitor of TNF-(alpha) USP4 USP5 and sentrin/SUMO-specific protease 1 (SENP1). Vialinin A (Terrestrin A) is able to be used for autoimmune diseases and cancer research [2] [3]. purity: 99%