Learn More
Medchemexpress LLC Vialinin A | 858134-23-3 | MFCD11113135 | 99.9% | 562.57 g·mol⁻1 | C34H26O8 | 1 MG
SDP
Catalog No. 5000244673
Encompass_Preferred
Vialinin A is a p-terphenyl natural product studied for its antioxidant and immunomodulatory activity. It is used in preclinical research as a potent inhibitor of tumor necrosis factor-alpha (TNF-α).
  • High purity suitable for analytical and biological studies.
  • Potent TNF-α inhibition demonstrated in assays.
  • Supplied as a solid for easy dissolution in common organic solvents.
  • Available in milligram quantities for screening and small-scale studies.
  • Molecular formula C34H26O8; molecular weight 562.57 g·mol⁻1.
Catalog No. 50-002-44673 Supplier Medchemexpress LLC Supplier No. HY1034351MG
Vialinin A is a p-terphenyl natural product studied for its antioxidant and immunomodulatory activity. It is used in preclinical research as a potent inhibitor of tumor necrosis factor-alpha (TNF-α).
  • High purity suitable for analytical and biological studies.
  • Potent TNF-α inhibition demonstrated in assays.
  • Supplied as a solid for easy dissolution in common organic solvents.
  • Available in milligram quantities for screening and small-scale studies.
  • Molecular formula C34H26O8; molecular weight 562.57 g·mol⁻1.