Learn More
Research Products International Corp Vial Rack, Polypropylene, for 5-8ml Vials, 90 Vial Capacity, 2 per Package
SDP
Catalog No. 50212470
Encompass
Heavy duty polypropylene racks have excellent chemical resistance and can be autoclaved. Ideal for use when processing samples or transporting samples.
Catalog No. 50-212-470 Supplier Research Products International Corp Supplier No. 224805
Heavy duty polypropylene racks have excellent chemical resistance and can be autoclaved. Ideal for use when processing samples or transporting samples.