Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Amber, 2ml, 10-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 5001452204
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass, amberized. Features a wide mouth opening for easy filling and ND10 threads. Amber vials are often used for light sensitive compounds and are considered low actinic.
Catalog No. 50-014-52204 Supplier Microsolv Technology Corporation Supplier No. 9503SE0AV
First hydrolytic, Class A, Borosilicate Glass, amberized. Features a wide mouth opening for easy filling and ND10 threads. Amber vials are often used for light sensitive compounds and are considered low actinic.