Learn More
Microsolv Technology Corporation Vials 10-425 Amber Wo 1.8Mlv
Catalog No. 5001452209
Encompass_Preferred
Vials, Screw Top, Glass. Amber, 2ml, with Write-On Patch and fill lines, 10-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52209 Supplier Microsolv Technology Corporation Supplier No. 9503SWAV
Vials, Screw Top, Glass. Amber, 2ml, with Write-On Patch and fill lines, 10-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. MicroSolv Brand. 100/PK.